missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLNK Antibody (125), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90140AF350
This item is not returnable.
View return policy
Description
BLNK Monoclonal specifically detects BLNK in Human samples. It is validated for ELISA.
Specifications
| BLNK | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 125 | |
| ELISA | |
| AGM4, B cell linker protein, BASH, B-cell adapter containing a SH2 domain protein, B-cell adapter containing a Src homology 2 domain protein, B-cell linker, B-cell linker protein, BLNK-s, Cytoplasmic adapter protein, LY57, MGC111051, SLP-65BLNK-S, SLP65Ly57, Src homology 2 domain-containing leukocyte protein of 65 kDa | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human BLNK (P08195-1; Met 1-Ser 456). | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 29760 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction