Learn More
BLMH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14941-1-AP-20UL
 View more versions of this product
Product Code. 16863864
  • £135.00 / 20µL
Description
The normal physiological role of BLM hydrolase is unknown, but it catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxamide bond of its B-aminoalaninamide moiety thus protecting normal and maligt cells from BLM toxicity.
Specifications
Specifications
BLMH
Polyclonal
Unconjugated
BLMH
BH, bleomycin hydrolase, BLM hydrolase, BLMH, BMH
Rabbit
Antigen Affinity Chromatography
RUO
104184, 287552, 642
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P70645, Q13867, Q8R016
Blmh
BLMH Fusion Protein Ag6888
20 μL
Primary
Human, Rat, Mouse