missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BLMH Monoclonal Antibody (760239)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561639

Product Code. 30535787

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

The normal physiological role of BLM hydrolase is unknown, but it catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxamide bond of its B-aminoalaninamide moiety thus protecting normal and malignant cells from BLM toxicity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BLMH
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q13867
BLMH
E. coli-derived recombinant human BLMH/Bleomycin Hydrolase, Ser2-Glu455.
100 μg
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
760239
Unconjugated
BLMH
AI035728; BH; Bleomycin hydrolase; BLM hydrolase; BLMH; BMH
Mouse
Protein A/G
RUO
642
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.