missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLMH/Bleomycin Hydrolase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88640
This item is not returnable.
View return policy
Description
BLMH/Bleomycin Hydrolase Polyclonal specifically detects BLMH/Bleomycin Hydrolase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BLMH/Bleomycin Hydrolase | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| BHBMHBLM hydrolase, bleomycin hydrolase, EC 3.4.22.40 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| BLMH | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RKEPEDGRLVQFLLMNPANDGGQWDMLVNIVEKYGVIPKKCFPESYTTEATRRMNDILNHKMREFCIRLRNLVHSGATKGEISATQDVMMEEIFRVVCICLGNP | |
| 0.1 mL | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 642 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction