missing translation for 'onlineSavingsMsg'
Learn More

Blk Antibody (7A12), Novus Biologicals™

Product Code. 18378467 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378467 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378467 Supplier Novus Biologicals Supplier No. H00000640M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Blk Monoclonal antibody specifically detects Blk in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blk
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 7A12
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry 1:10 to 1:1000, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH07371
Gene Alias B lymphocyte kinase, B lymphoid tyrosine kinase, BLK nonreceptor tyrosine kinase, EC 2.7.10.2, MGC10442, MODY11, p55-Blk, tyrosine-protein kinase Blk
Host Species Mouse
Immunogen BLK (AAH07371, 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGLVSSKKPDKEKPIKEKDKGQWSPLKVSAQDKDAPPLPPLVVFNHLTPPPPDEHLDEDKHFVVALYDYTAMNDRDLQMLKGEKLQVLKG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 640
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.