missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLIMP1/PRDM1 Antibody (646702) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB36081JF585
This item is not returnable.
View return policy
Description
BLIMP1/PRDM1 Monoclonal antibody specifically detects BLIMP1/PRDM1 in Human samples. It is validated for Western Blot,Simple Western,Flow Cytometry (Intracellular Staining)
Specifications
| BLIMP1/PRDM1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PR-domain zinc finger protein 1 | |
| E. coli-derived recombinant human BLIMP1/PRDM1, Lys667-Cys789, Accession # O75626 | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Cancer, Chromatin Research, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Zinc Finger | |
| 639 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Simple Western, Flow Cytometry (Intracellular Staining) | |
| 646702 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction