missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Blimp-1 Monoclonal Antibody (5E7), APC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£161.00 - £357.00
Specifications
| Antigen | Blimp-1 |
|---|---|
| Clone | 5E7 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17111682
|
Invitrogen™
17-9850-80 |
25 μg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17121682
![]() |
Invitrogen™
17-9850-82 |
100 μg |
£357.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Blimp-1 (B lymphocyte maturation protein-1) is a nuclear protein with five zinc fingers in its carboxy terminus. It is a transcriptional repressor that is both required and sufficient to trigger terminal differentiation of B lymphocytes and monocyte/macrophages. Blimp-1 is expressed in, and may be required for terminally differentiated epithelial cells, retinal neurons and other lineage cells. It has also been implicated in the repression of beta-interferon (IFN-beta) and c-myc gene expression.Specifications
| Blimp-1 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| B lymphocyte-induced maturation protein 1; b2b1765Clo; Beta-interferon gene positive regulatory domain I-binding factor; beta-interferon gene positive-regulatory domain I binding factor; BLIMP 1; Blimp1; BLIMP-1; B-lymphocyte-induced maturation protein 1; MGC118922; MGC118923; MGC118924; MGC118925; OTTHUMP00000016918; OTTHUMP00000202173; Positive regulatory domain I-binding factor 1; PR domain 1; PR domain containing 1, with ZNF domain; PR domain zinc finger protein 1; PR domain-containing protein 1; PR/SET domain 1; PRDI BF1; PRDI-BF1; PRDI-binding factor-1; Prdm1; RP1-134E15.1; ZNFPR1A1 | |
| PRDM1 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| PRDM1 |
| 5E7 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Mouse | |
| Q60636 | |
| 12142 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title