missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blimp-1 Rat anti-Human,Mouse, PE-CF594, Clone: 6D3, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 565274
This item is not returnable.
View return policy
Description
Blimp-1 Monoclonal antibody specifically detects Blimp-1 in Human,Mouse samples. It is validated for Flow Cytometry.
Specifications
| Blimp-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 6D3 | |
| PE-CF594 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction