missing translation for 'onlineSavingsMsg'
Learn More

Blimp-1 Rat anti-Human, Mouse, Brilliant Violet 421, Clone: 6D3, BD Horizon™

Product Code. 15884199
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15884199 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15884199 Supplier BD Biosciences Supplier No. 565276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Blimp-1 Monoclonal antibody specifically detects Blimp-1 in Human, Mouse samples. It is validated for Immunocytochemistry, Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blimp-1
Applications Immunocytochemistry, Flow Cytometry
Classification Monoclonal
Clone 6D3
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Host Species Rat
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Blimp-1
Target Species Human, Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.