Learn More
Invitrogen™ Blimp-1 Monoclonal Antibody (3H2-E8), DyLight™ 488

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516115
Description
In Western Blot, this antibody recognizes a band at ~98 kDa and may recognize one at ~80 kDa (the beta form). Antigen retrieval is recommended (EDTA buffer, microwave) prior to IHC on paraffin tissues. This antibody does not immunoprecipitate.For IHC and ICC/IF a dilution of 1:200 is recommended with tyramide amplification; this antibody demonstrates nuclear staining. Suggested positive control: P3X (murine plasmocytoma) cell lysates.
Blimp-1 (B lymphocyte maturation protein-1) is a nuclear protein with five zinc fingers in its carboxy terminus. It is a transcriptional repressor that is both required and sufficient to trigger terminal differentiation of B lymphocytes and monocyte/macrophages. Blimp-1 is expressed in, and may be required for terminally differentiated epithelial cells, retinal neurons and other lineage cells. It has also been implicated in the repression of beta-interferon (IFN-beta) and c-myc gene expression.
Specifications
| Blimp-1 | |
| Monoclonal | |
| 0.67 mg/mL | |
| 50mM sodium borate with 0.05% sodium azide | |
| O75626, Q60636 | |
| PRDM1 | |
| A fragment of mouse Blimp-1 corresponding to residues 199-409. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Pig | |
| Antibody | |
| IgG1 |
| ChIP Assay, ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 3H2-E8 | |
| DyLight 488 | |
| PRDM1 | |
| B lymphocyte-induced maturation protein 1; b2b1765Clo; Beta-interferon gene positive regulatory domain I-binding factor; beta-interferon gene positive-regulatory domain I binding factor; BLIMP 1; Blimp1; BLIMP-1; B-lymphocyte-induced maturation protein 1; MGC118922; MGC118923; MGC118924; MGC118925; OTTHUMP00000016918; OTTHUMP00000202173; Positive regulatory domain I-binding factor 1; PR domain 1; PR domain containing 1, with ZNF domain; PR domain zinc finger protein 1; PR domain-containing protein 1; PR/SET domain 1; PRDI BF1; PRDI-BF1; PRDI-binding factor-1; Prdm1; RP1-134E15.1; ZNFPR1A1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100154284, 12142, 639 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.