missing translation for 'onlineSavingsMsg'
Learn More

Bleomycin sulfate

Product Code. p-7041921
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483439 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15483439 Supplier Thermo Scientific Alfa Aesar Supplier No. J60727.MA
Add to basket
This item is not returnable. View return policy

Bleomycin sulfate, CAS # 9041-93-4, is a glycopeptide compound with antibiotic activity isolated from Streptomyces verticillus. It also exhibits anticancer activity. | CAS: 9041-93-4 | C55H85N17O25S4 | 1512.619 g/mol

Bleomycin sulfate is used as a glycopeptide antibiotic. It is capable of inducing oxidative cleavage of the DNA backbone, leading to apoptosis and also degrades RNA, and has been reported to display selectivity for RNA over DNA. Bleomycin acts as a sclerosing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Bleomycin sulfate was initially isolated from Streptomyces verticillus. It is a glycopeptide compound with antibiotic activity
  • It has the ability to induce oxidative cleavage of the DNA backbone, leading to apoptosis
  • Bleomycin sulfate can form a complex with iron that reduces oxygen to superoxide and hydroxyl radicals. These reactive oxygen species can cause single- and double-stranded breaks in DNA as well as promote carbohydrate oxidation, lipid peroxidation, and alterations in prostaglandin synthesis and degradation

Application

  • It can be used to inhibit DNA metabolism and slow or stop cell growth. This property explains its anticancer activity
  • Bleomycin sulfate can also be used to degrade RNA as it is more selective to RNA than DNA
  • Local injections of bleomycin sulfate in mouse models can induce sclerotic skin mimicking scleroderma
  • In laboratory research, bleomycin is used to induce pulmonary fibrosis in mice
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-93-4
Molecular Formula C55H85N17O25S4
Molecular Weight (g/mol) 1512.619
MDL Number MFCD00070310
InChI Key WUIABRMSWOKTOF-UHFFFAOYSA-N
PubChem CID 131664136
IUPAC Name [2-[2-[2-[[6-amino-2-[3-amino-1-[(2,3-diamino-3-oxopropyl)amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[[5-[[1-[2-[4-[4-(3-dimethylsulfoniopropylcarbamoyl)-1,3-thiazol-2-yl]-1,3-thiazol-2-yl]ethylamino]-3-hydroxy-1-oxobutan-2-yl]amino]-3-hyd
SMILES CC1=C(N=C(N=C1N)C(CC(=O)N)NCC(C(=O)N)N)C(=O)NC(C(C2=CN=CN2)OC3C(C(C(C(O3)CO)O)O)OC4C(C(C(C(O4)CO)O)OC(=N)[O-])O)C(=O)NC(C)C(C(C)C(=O)NC(C(C)O)C(=O)NCCC5=NC(=CS5)C6=NC(=CS6)C(=O)NCCC[S+](C)C)O.OS(=O)(=O)O

Specifications

Type Bleomycin Sulfate
Physical Form Powder
Color White
Quantity 10 mg
Merck Index 14,1318
Solubility Information Soluble in water or DMSO
Chemical Name or Material Bleomycin sulfate
compliance-icons
Product Identifier
  • Bleomycin sulfate
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H340-May cause genetic defects.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Bleomycin, sulfate (salt)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.