missing translation for 'onlineSavingsMsg'
Learn More

Bleomycin Sulfate (A mixture of bleomycin sulfate salts) >90%, TRC

Product Code. 30041192
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041192 50 mg 50mg
30041150 5 mg 5mg
30041143 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041192 Supplier Toronto Research Chemicals Supplier No. TRCB59580050MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-93-4
Molecular Formula C55 H84 N17 O21 S3 . C55 H84 N20 O21 S2 . H O4 S . H2 O4 S
Molecular Weight (g/mol) 3036.21
Synonym Bleomycin, sulfate (salt), Blenoxane, Bleomycin sulfate, Blexane
IUPAC Name [(2R,3S,4S,5R,6R)-2-[(2R,3S,4S,5S,6S)-2-[(1R,2S)-2-[[6-amino-2-[(1S)-3-amino-1-[[(2S)-2,3-diamino-3-oxopropyl]amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[[(2R,3S,4S)-5-[[(2S,3R)-1-[2-[4-[4-(4-carbamimidamidobutylcarbamoyl)-1,3-thiazol-2-yl]-1,3-thiazol-2-yl]ethylamino]-3-hydroxy-1-oxobutan-2-yl]amino]-3-hydroxy-4-methyl-5-oxopentan-2-yl]amino]-1-(1H-imidazol-5-yl)-3-oxopropoxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]oxy-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl] carbamate;3-[[2-[2-[2-[[(2S,3R)-2-[[(2S,3S,4R)-4-[[(2S,3R)-2-[[6-amino-2-[(1S)-3-amino-1-[[(2S)-2,3-diamino-3-oxopropyl]amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[(2R,3S,4S,5S,6S)-3-[(2R,3S,4S,5R,6R)-4-carbamoyloxy-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-(1H-imidazol-5-yl)propanoyl]amino]-3-hydroxy-2-methylpentanoyl]amino]-3-hydroxybutanoyl]amino]ethyl]-1,3-thiazol-4-yl]-1,3-thiazole-4-carbonyl]amino]propyl-dimethylsulfanium;hydrogen sulfate;sulfuric acid
SMILES C[C@@H](O)[C@H](NC(=O)[C@@H](C)[C@H](O)[C@@H](C)NC(=O)[C@@H](NC(=O)c1nc(nc(N)c1C)[C@H](CC(=O)N)NC[C@H](N)C(=O)N)[C@@H](O[C@@H]2O[C@@H](CO)[C@@H](O)[C@H](O)[C@@H]2O[C@H]3O[C@H](CO)[C@@H](O)[C@H](OC(=O)N)[C@@H]3O)c4cnc[nH]4)C(=O)NCCc5nc(cs5)c6nc(cs6)C(=O)NCCCCNC(=N)N.C[C@@H](O)[C@H](NC(=O)[C@@H](C)[C@H](O)[C@@H](C)NC(=O)[C@@H](NC(=O)c7nc(nc(N)c7C)[C@H](CC(=O)N)NC[C@H](N)C(=O)N)[C@@H](O[C@@H]8O[C@@H](CO)[C@@H](O)[C@H](O)[C@@H]8O[C@H]9O[C@H](CO)[C@@H](O)[C@H](OC(=O)N)[C@@H]9O)c%10cnc[nH]%10)C(=O)NCCc%11nc(cs%11)c%12nc(cs%12)C(=O)NCCC[S+](C)C.OS(=O)(=O)O.OS(=O)(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C55H84N20O21S2.C55H83N17O21S3.2H2O4S/c1-19-32(72-45(75-43(19)58)24(11-30(57)79)67-12-23(56)44(59)85)49(89)74-34(40(25-13-63-18-68-25)94-53-42(38(83)36(81)28(14-76)93-53)95-52-39(84)41(96-55(62)91)37(82)29(15-77)92-52)50(90)69-21(3)35(80)20(2)46(86)73-33(22(4)78)48(88)65-10-7-31-70-27(17-97-31)51-71-26(16-98-51)47(87)64-8-5-6-9-66-54(60)61;1-20-33(69-46(72-44(20)58)25(12-31(57)76)64-13-24(56)45(59)82)50(86)71-35(41(26-14-61-19-65-26)91-54-43(39(80)37(78)29(15-73)90-54)92-53-40(81)42(93-55(60)88)38(79)30(16-74)89-53)51(87)66-22(3)36(77)21(2)47(83)70-34(23(4)75)49(85)63-10-8-32-67-28(18-94-32)52-68-27(17-95-52)48(84)62-9-7-11-96(5)6;2*1-5(2,3)4/h13,16-18,20-24,28-29,33-42,52-53,67,76-78,80-84H,5-12,14-15,56H2,1-4H3,(H2,57,79)(H2,59,85)(H2,62,91)(H,63,68)(H,64,87)(H,65,88)(H,69,90)(H,73,86)(H,74,89)(H2,58,72,75)(H4,60,61,66);14,17-19,21-25,29-30,34-43,53-54,64,73-75,77-81H,7-13,15-16,56H2,1-6H3,(H13-,57,58,59,60,61,62,63,65,66,69,70,71,72,76,82,83,84,85,86,87,88);2*(H2,1,2,3,4)/t20-,21+,22+,23-,24-,28-,29+,33-,34-,35-,36+,37+,38-,39-,40-,41-,42-,52+,53-;21-,22+,23+,24-,25-,29-,30+,34-,35-,36-,37+,38+,39-,40-,41-,42-,43-,53+,54-;;/m00../s1
Formula Weight 3034.002
Chemical Name or Material Bleomycin Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.