missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blasticidin-S, hydrochloride, 99.2%, For HPLC analysis, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
Specifications
Specifications
| Type | Blasticidin S Hydrochloride |
| Physical Form | Powder |
| Color | White |
| Quantity | 100 mg |
| Assay Percent Range | 0.992 |
| Formula Weight | 458.9 |
| Chemical Name or Material | Blasticidin-S hydrochloride, For HPLC analysis |
Safety and Handling
Product Identifier
- Blasticidin S Hydrochlorid
- Danger
- Acute toxicity Category 1
- H300-Fatal if swallowed.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction