missing translation for 'onlineSavingsMsg'
Learn More

Blasticidin-S, hydrochloride, 99.2%, For HPLC analysis, MP Biomedicals™

Product Code. 11417731 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 11417731

Brand: MP Biomedicals 0215047725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3-9-3513
Molecular Formula C17H27ClN8O5
Molecular Weight (g/mol) 458.90
InChI Key YQXYQOXRCNEATG-UHFFFAOYNA-N
Synonym BLA-S, Cytovirin
SMILES Cl.CN(CCC(N)CC(=O)NC1C=CC(OC1C(O)=O)N1C=CC(N)=NC1=O)C(N)=N

Specifications

Type Blasticidin S Hydrochloride
Physical Form Powder
Color White
Quantity 25 mg
Assay Percent Range 0.992
Formula Weight 458.9
Chemical Name or Material Blasticidin-S hydrochloride, For HPLC analysis
compliance-icons
Product Identifier
  • Blasticidin S Hydrochlorid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.