missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BLAP75 Polyclonal specifically detects BLAP75 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | BLAP75 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000-1:10000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1-4 ug/mg of lysate, Immunohistochemistry-Paraffin 1:100-1:500 |
| Formulation | TBS and 0.1% BSA with 0.09% Sodium Azide |
| Gene Accession No. | NP_079221.1 |
| Gene Alias | 75 kDa, BLM-associated protein 75 kDa, BLM-associated protein of 75 kDa, chromosome 9 open reading frame 76, FAAP75, FLJ12888, homolog of yeast RecQ-mediated genome instability 1 (RMI1), recQ-mediated genome instability protein 1, RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae), RMI2 |
| Gene Symbols | RMI1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?