missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLAME/SLAMF8 Rabbit anti-Mouse, Clone: 210, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £429.00
Specifications
| Antigen | BLAME/SLAMF8 |
|---|---|
| Clone | 210 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261408
|
Novus Biologicals
NBP2-90656-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18266646
|
Novus Biologicals
NBP2-90656-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BLAME/SLAMF8 Monoclonal specifically detects BLAME/SLAMF8 in Mouse samples. It is validated for ELISA.Specifications
| BLAME/SLAMF8 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 56833 | |
| IgG | |
| Protein A purified |
| 210 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| B Lymphocyte Activator Macrophage Expressed, BCM-Like Membrane Protein, B-Lymphocyte Activator Macrophage Expressed, CD353, CD353 Antigen, SBBI42, SLAM Family Member 8, SLAMF8 | |
| Recombinant Mouse BLAME/SLAMF8 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title