missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Blade Holder Packages for the Cryotome™ FSE Cryostat, Low-Profile Cryotome Blade Holder

Product Code. p-4564342 Shop All Epredia Products
Click to view available options
Product Type:
Low Profile Blade Holder
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12111840

Brand: Epredia™ A78910032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hold blades for the Cryotome™ FSE Cryostat with these packages.

Universal Blade Holder:
  • Accepts standard C- or D-profile knife up to 220mm; 120mm is standard length
  • Use with standard steel knives or with disposable blade holder
  • Use in conjunction with disposable blade holders (Part Nos. 40 or 47)
Low-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder for use with low-profile disposable blades
High-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder will accept either standard or heavy-duty high-profile blades, providing a wide range of tissue sectioning options
TRUSTED_SUSTAINABILITY

Specifications

Product Type Low Profile Blade Holder
For Use With (Equipment) FSE/FE/SME/E
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.