Learn More
Epredia™ Blade Carriers
Product Code. 12023079 Shop All Epredia Products
Click to view available options
Product Type:
Blade holder insert
SN Type Knife carrier base
Unit Size:
Each
Product Code. 12023079
Brand: Epredia™ 565060
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Insert a variety of Epredia™ blades in Epredia™ Blade Carriers.
Updated Knife Carriers:
  • Blade angle and blade adjustment levers are removable to help protect user settings
  • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder
  • Compatible only with newer HM 325, HM 355S, and HM 340E models which have serial numbers above 34000
Standard Knife Carriers:
  • Integrated knife guard with clearance angle adjustment.
  • Precision guidance
  • Complete with knife carrier base
  • Backlighting system adaptable
  • Laterally movable to accommodate conventional knives, tungsten carbide knives, and disposable blade inserts
Blade Holder Insert:
  • For low-profile blades to be inserted in the standard knife carriers N and SN (includes Allen key)
Knife Carrier Base:
  • Carrier base with clamping device
  • Use along with the upper parts of the knife carriers N, SN, NR, ER, C, SL
TRUSTED_SUSTAINABILITY
Specifications
Product Type SN Type Knife carrier base
For Use With (Equipment) Dovetail Style Unit