Learn More
Epredia™ Blade Carriers
Insert a variety of Epredia™ blades in Epredia™ Blade Carriers.
Brand: Epredia™ 705420
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Updated Knife Carriers:
- Blade angle and blade adjustment levers are removable to help protect user settings
- Concentric lever locks onto the dovetail to help reduce accidental movement of the holder
- Compatible only with newer HM 325, HM 355S, and HM 340E models which have serial numbers above 34000
Standard Knife Carriers:
- Integrated knife guard with clearance angle adjustment.
- Precision guidance
- Complete with knife carrier base
- Backlighting system adaptable
- Laterally movable to accommodate conventional knives, tungsten carbide knives, and disposable blade inserts
Blade Holder Insert:
- For low-profile blades to be inserted in the standard knife carriers N and SN (includes Allen key)
Knife Carrier Base:
- Carrier base with clamping device
- Use along with the upper parts of the knife carriers N, SN, NR, ER, C, SL
Specifications
Blade holder insert | |
Insert |