Learn More
Motic™ Black/White Stage Plate, 80mm
Select a microscope stage plate, based on sample type, that provides you with the best background for viewing. Motic™ Black Stage Plate, 80mm is one of several possible 80mm stage plates available from Motic™.
Brand: Motic™ 1101007400061
Additional Details : Weight : 0.01000kg
Description
- Replaceable, optional stage plate insert.
- Provides the most appropriate background for viewing.
- Reversible, dual color plate.
Specifications
Reversible Black/White Stage Plate | |
80 mm | |
SMZ-140, SFC-11, SMZ-161, DM-143 Microscopes |