missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Black Urea Screw Caps with Wood Pulp Faced PVDC Liner
Click to view available options
Neck Finish:
100/R3
22/R3
24/R3
28/R3
31/R6
33/R3
38/R3
48/R3
51/R3
58/R3
63/R3
83/R3
Unit Size:
Pack of 50
Description
- Black urea plastic
- Includes fitted wood pulp faced PVDC liner
- Supplied in packs of 50
Specifications
Specifications
| Color | Black |
| Material | Urea |
| Quantity | 50/PK |
| Liner Material | Wood pulp faced PVDC |
| Neck Finish | 63/R3 |
| Product Type | Screw Cap |
| For Use With (Equipment) | Wide mouth bottles and jars |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction