missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Black Urea Screw Caps with Polythene Cone Liner
Description
- For use with Winchester bottles
- Constructed of black plastic urea
- Polythene cone liner
- Ridged for better grip
Specifications
Closure Size | 31/R3 |
Closure Type | Cone-lined Screw Cap |
For Use With (Equipment) | Winchester bottles |
Color | Black |
Material | Urea |
Quantity | 50/PK |
Liner Material | Polythene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'