missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Black Urea Screw Caps with Polythene Cone Liner
Click to view available options
Neck Finish:
20/R3
22/R3
24/R3
28/R3
31/R3
Unit Size:
Pack of 50
Description
- For use with Winchester bottles
- Constructed of black plastic urea
- Polythene cone liner
- Ridged for better grip
Specifications
Specifications
| Color | Black |
| Material | Urea |
| Quantity | 50/PK |
| Liner Material | Polythene |
| Neck Finish | 22/R3 |
| Product Type | Cone-lined Screw Cap |
| For Use With (Equipment) | Winchester bottles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction