Learn More
PSF Progressive Safety™ Black Rubber Waterproof Safety Boot
Product Code. 12974085
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
1 pair
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12974085 6 1 pair
12984085 7 1 pair
12994085 8 1 pair
12904095 9 -
12944085 10 1 pair
12954085 11 1 pair
12964085 12 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12974085 Supplier PSF Progressive Safety™ Supplier No. STOPRAIN6
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PVC & nitrile rubber upper, phthalate free
  • PVC & nitrile rubber upper, phthalate free
  • 100% waterproof
  • Double stitched seams to upper
  • Four D-ring fasten
  • Antistatic
  • Steel toecap and midsole
  • Large external toe and heel protection
  • Classed as hybrid PPE
  • Certified against some chemicals, please enquire
  • SRA slip resistant
Specifications
Size 6