Learn More
Bohlender™ Black PPS/PTFE Screw Cap
Product Code. 13296869
Click to view available options
:
For Use With: 1.6 to 10mm O.D. tubing
Product Code. 13296869
Brand: Bohlender™ D78416
Only null left
Add to basket
Add to basket
For connecting or inserting tubing, tubes or probes. BOLA screw caps feature universal chemical resistance.
  • Black screw cap made of PPS
  • Movable reduction body made of PTFE with O-ring made of FKM for transition to GL threads
  • ody can be moved independently from the screw cap
  • Completely assembled reduction can be removed and fixed on another vessel without the risk of disarranging the tubing or cable
  • Product is only exposed to PTFE