Learn More
Parisienne de Verrerie™ Black Phenolic Resin Screw Caps
Providing secure closure for a variety of bottle neck sizes. Parisienne de Verrerie™ Black Phenloic Resin Screw Caps are durable and resilient.
Brand:  Parisienne de Verrerie™ P08028
Product Code. 11703853
  • ONLINE EXCLUSIVE PRICE
    £170.00
    £138.00 / Pack of 100
    Save £32.00
    18% Off
Only null left
In Stock
Add to basket
Description
Description
  • Polyethylene-coated chipboard liners temperature resistant up to 80°C
  • Rubber liners fully autoclavable
  • Polyethylene foam liners temperature resistant up to 60°C
  • Butyl-PTFE liners fully autoclavable