missing translation for 'onlineSavingsMsg'
Learn More

PSF Progressive Safety™ Black Leather D Ring Chukka Boot

Product Code. 15553621
Click to view available options
Size:
7
9
10
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 15553621

Brand: PSF Progressive Safety™ 1007

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tough footwear for tough work, resistant to oil and static.

  • All safety footwear is designed with both comfort and durability in mind
  • Conforms to EN ISO 20345:2011 for 200 joules protection
  • Dual density polyurethane sole
  • 4 D-ring fasten with black round laces
  • Breathable lining
  • Padded collar and bellows tongue
  • Slip-resistant sole

Specifications

Certifications/Compliance EN ISO 20345:2011
Color Black
Cuff Style Padded
Design D-ring
For Use With (Application) Tough work conditions
Material Leather
Size 7
Sole Material Dual density polyurethane
Style Chukka boot
Resistant To Oil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.