Learn More
Macherey Nagel Black Filter Paper MN 220
Used to identify small quantities of light precipitates.
Brand:  Macherey Nagel 409007
Additional Details : Weight : 0.10000kg
Product Code. 10042651
  • £14.70 / Pack of 100
Estimated Shipment: 27-07-2022
to see stock.
Add to basket
Description
Description
  • Dyed black with a sulfur dye
  • Used to identify small quantities of light precipitates such as the detection of fluorine or silicone
Specifications
Specifications
85 g/m2
100/Pk.
70 mm
MN 220
0.17 mm