missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BL 1249
GREENER_CHOICE

Product Code. 18075382
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18075382

Brand: Tocris Bioscience™ 3797/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Putative K2P2.1 (TREK-1) channel opener

Putative potassium channel activator; thought to act as a K2P2.1 (TREK-1) channel opener. Exhibits selectivity for bladder over vascular tissue in vitro and in vivo (EC50 values are 1.26 and 21.0 μM for cultured bladder and aortic tissues respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18200-13-0
Molecular Formula C17H17N5

Specifications

Quantity 10 mg
Formula Weight 291.35
Purity >99%
Molecular Weight (g/mol) 291.358
Chemical Name or Material BL 1249
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.