missing translation for 'onlineSavingsMsg'
Learn More

K-604 dihydrochloride, MedChemExpress

Product Code. 16415737
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16415737

Brand: Medchem Express HY100400A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress K-604 dihydrochloride is a potent and selective acyl-CoA:cholesterol acyltransferase 1 (ACAT-1) inhibitor with an IC50 of 0.45±0.06 μM.

K-604 dihydrochloride is a potent and selective acyl-CoA:cholesterol acyltransferase 1 (ACAT-1) inhibitor with an IC50 of 0.45±0.06 μM.

Specifications

Chemical Name or Material K-604 dihydrochloride
CAS 217094-32-1
Color White
Purity Grade Notes Research
Molecular Formula C23H32Cl2N6OS3
Quantity 5 mg
Solubility Information H2O : 100 mg/mL (173.72 mM; Need ultrasonic) ∣DMSO : 62.5 mg/mL (108.57 mM; Need ultrasonic)
SMILES O=C(NC1=C(SC)C=C(C)N=C1SC)CN2CCN(CCSC3=NC4=CC=CC=C4N3)CC2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 575.64
Formula Weight 575.64
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.