missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies BIX01294 (Trihydrochloride Hydrate)

Product Code. 17180012
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 17180012

Brand: STEMCELL Technologies 72042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Epigenetic modifier; Inhibits G9a histone methyltransferase

  • BIX01294 is a very specific inhibitor of the G9a (IC50 = 1.9 μM) and G9a-like (GLP, IC50 = 0.7 μM) histone methyltransferases
  • These methyltransferases target the lysine 9 position on histone 3 (H3K9)
  • BIX01294 inhibits G9a and GLP by occupying the histone binding site, preventing interaction with histones (Chang et al.), Kubicek et al.) This product is supplied as the trihydrochloride hydrate form of the molecule
  • Enhances reprogramming of mouse embryonic fibroblasts or fetal neural progenitor cells to induced pluripotent stem cells without using c-Myc and SOX2 (Shi et al. 2008a, Shi et al. 2008b)

Specifications

CAS 1808255-64-2
Quantity 1 mg
Molecular Formula C28H38N6O2 · 3HCl [XH2O]
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.