missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIX-01294, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BIX-01294 is a reversible and highly selective G9a and GLP Histone Methyltransferase inhibitor, with IC50s of of 1.7 μM and 0.9 μM, respectively. BIX-01294 inhibits G9a/GLP by competing for binding with the amino acids N-terminal of the substrate lysine residue. BIX-01294, a (1H-1,4-diazepin-1-yl)-quinazolin-4-yl amine derivative, induces necroptosis and autophagy. BIX-01294 has antitumor activity in recurrent tumor cells.
Specifications
Specifications
| Chemical Name or Material | BIX-01294 |
| CAS | 935693-62-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H38N6O2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 110 mg/mL (224.20 mM) ∣H2O : 1 mg/mL (2.04 mM; Need ultrasonic) |
| SMILES | COC1=C(OC)C=C(C(NC2CCN(CC3=CC=CC=C3)CC2)=NC(N4CCN(C)CCC4)=N5)C5=C1 |
| Molecular Weight (g/mol) | 490.64 |
| Formula Weight | 490.64 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction