Learn More
BIVM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25432-1-AP-150UL
 View more versions of this product
Product Code. 16874005
  • £349.00 / 150µL
Description
BIVM has been identified using an electronic search based on the conservation of short sequence motifs within the variable region of immunoglobulin (Ig) genes. It is tightly linked (41 bp) and in the opposite transcriptional orientation to MGC5302 (also known as KDEL1 and EP58) in human. BIVM has two isoforms with MW 57 kDa and 32 kDa.
Specifications
Specifications
BIVM
Polyclonal
Unconjugated
BIVM
BIVM
Rabbit
Antigen Affinity Chromatography
RUO
54841
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86UB2
BIVM
BIVM Fusion Protein Ag22087
150 μL