missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BIVM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522115
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Dog (100%), Pig (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BIVM has been identified using an electronic search based on the conservation of short sequence motifs within the variable region of immunoglobulin (Ig) genes. It is tightly linked (41 bp) and in the opposite transcriptional orientation to MGC5302 (also known as KDEL1 and EP58) in human. BIVM has two isoforms with MW 57 kDa and 32 kDa.
Specifications
| BIVM | |
| Polyclonal | |
| Unconjugated | |
| BIVM | |
| AI854267; basic immunoglobulin-like variable motif-containing protein; basic immunoglobulin-like variable motif-containing protein-like; basic, immunoglobulin-like variable motif containing; BIVM; LOC102548228; RP11-484I6.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54841 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q86UB2 | |
| BIVM | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human BIVM. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction