missing translation for 'onlineSavingsMsg'
Learn More

Bivalirudin trifluoroacetate, Thermo Scientific™

Product Code. 17595565 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17595565 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17595565 Supplier Thermo Scientific Acros Supplier No. 469101000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Purity ≥97.5 %
CAS 1191386-55-6
Color White
Molecular Formula C98H138N24O33
Molecular Weight (g/mol) 2294.33, 2180.32
InChI Key OIRCOABEOLEUMC-UHFFFAOYNA-N
SMILES CCC(C)C(NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CC1=CC=CC=C1)NC(=O)C(CC(O)=O)NC(=O)CNC(=O)C(CC(N)=O)NC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)C1CCCN1C(=O)C(CCCNC(N)=N)NC(=O)C1CCCN1C(=O)C(N)CC1=CC=CC=C1)C(=O)N1CCCC1C(=O)NC(CCC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CC1=CC=C(O)C=C1)C(=O)NC(CC(C)C)C(O)=O
IUPAC Name 4-[2-(2-{2-[2-(2-{2-[2-(2-{[1-(2-{[1-(2-amino-3-phenylpropanoyl)pyrrolidin-2-yl]formamido}-5-carbamimidamidopentanoyl)pyrrolidin-2-yl]formamido}acetamido)acetamido]acetamido}acetamido)-3-carbamoylpropanamido]acetamido}-3-carboxypropanamido)-3-phenylpropanamido]-4-({3-carboxy-1-[(1-{2-[(3-carboxy-1-{[3-carboxy-1-({1-[(1-carboxy-3-methylbutyl)carbamoyl]-2-(4-hydroxyphenyl)ethyl}carbamoyl)propyl]carbamoyl}propyl)carbamoyl]pyrrolidin-1-yl}-3-methyl-1-oxopentan-2-yl)carbamoyl]propyl}carbamoyl)butanoic acid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.