missing translation for 'onlineSavingsMsg'
Learn More

Biuret Reagent - British Pharmacopoeia, Fisherbrand™

Product Code. 30695061 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30695061 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30695061 Supplier Fisher Chemical Supplier No. BPR 051

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents for testing and monographs, produced under a GMP-compliant ISO 9001 quality management system.

    This Pharmacopoeia reagent is manufactured :
  • Using pharmacopoeia reagent quality components
  • Water meeting the relevant pharmacopoeia monograph requirement for Purified Water
  • Supplied with a Certificate of Analysis for Performance Test
  • Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO 17034

Hazard Statement: May be flammable or oxidizing; may cause severe skin burns and serious eye damage or irritation; may cause respiratory irritation, drowsiness or dizziness; may be toxic if swallowed, in contact with skin, or if inhaled; may cause cancer, genetic defects, reproductive toxicity or developmental harm; may be harmful to aquatic life with long-lasting effects; may be corrosive to metals.

Specifications

Quantity 1000 mL
compliance-icons
Product Identifier
  • British Pharmacopoeia Reagent - Biuret Reagent
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide, copper sulphate, pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.