missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CellPath BiTran™ High Visibility Barrier Film Specimen Bag
Designed for transporting wet tissue from one place to another, and for the short to long-term storage of tissue.
£443.00 - £467.00
Specifications
Product Type | Specimen Bag |
---|---|
Closure Type | Zip Seal |
For Use With (Application) | For transporting wet tissue, short to long-term storage of tissue |
Material | High-visibility Barrier Film |
Quantity | 250 Pack |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
12320649
|
CellPath
WAA-0400-12A |
180 x 200mm (7 x 8 in.) |
£467.00
Pack of 250 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
12310649
|
CellPath
WAA-0300-12A |
150 x 150mm (6 x 6 in.) |
£443.00
Pack of 250 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
- Re-sealable specimen bags
- Manufactured from a special barrier film with a unique double zip lock closure
- Barrier film is impermeable to formalin vapour, preventing hazardous vapour polluting the laboratory once the bag is sealed
- Air is expelled from the bag by squeezing prior to closure
- Double zip lock seal provides a leak proof closure that can be easily opened and closed as many times as required allowing the specimen to be re-examined whenever necessary
Specifications
Specimen Bag | |
For transporting wet tissue, short to long-term storage of tissue | |
250 Pack |
Zip Seal | |
High-visibility Barrier Film |