missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CellPath BiTran™ High Visibility Barrier Film Specimen Bag
Click to view available options
Dimensions (L x W):
150 x 150mm (6 x 6 in.)
180 x 200mm (7 x 8 in.)
Unit Size:
Pack of 250
Description
- Re-sealable specimen bags
- Manufactured from a special barrier film with a unique double zip lock closure
- Barrier film is impermeable to formalin vapour, preventing hazardous vapour polluting the laboratory once the bag is sealed
- Air is expelled from the bag by squeezing prior to closure
- Double zip lock seal provides a leak proof closure that can be easily opened and closed as many times as required allowing the specimen to be re-examined whenever necessary
Specifications
Specifications
| Closure Type | Zip Seal |
| For Use With (Application) | For transporting wet tissue, short to long-term storage of tissue |
| Material | High-visibility Barrier Film |
| Quantity | 250 Pack |
| Dimensions (L x W) | 180 x 200mm (7 x 8 in.) |
| Product Type | Specimen Bag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction