Learn More
CellPath BiTran™ High Visibility Barrier Film Specimen Bag
Product Code. 12340649
Click to view available options
Dimensions (L x W):
300 x 300mm
Unit Size:
Pack of 250
Product Code. 12340649
Brand: CellPath WAA060012A
Only null left
Add to basket
Add to basket
Leak proof bags for the safe transportation of specimens
  • For transporting wet tissue from one place to another
  • Available in six sizes
  • Barrier film impermeable to formalin vapour
  • Double zip lock seal provides leak proof closure
Specifications
Closure Type Zip Seal
For Use With (Application) For transporting wet tissue, short to long-term storage of tissue
Material High-visibility Barrier Film
Quantity 250 Pack
Dimensions (L x W) 300 x 300mm
Product Type Specimen Bag