Learn More
CellPath BiTran™ High Visibility Barrier Film Specimen Bag
Click to view available options
Dimensions (L x W):
230 x 300mm
Unit Size:
Pack of 250
Description
- For transporting wet tissue from one place to another
- Available in six sizes
- Barrier film impermeable to formalin vapour
- Double zip lock seal provides leak proof closure
Specifications
Specifications
| Closure Type | Zip Seal |
| For Use With (Application) | For transporting wet tissue, short to long-term storage of tissue |
| Material | High-visibility Barrier Film |
| Quantity | 250 Pack |
| Dimensions (L x W) | 230 x 300mm |
| Product Type | Specimen Bag |