missing translation for 'onlineSavingsMsg'
Learn More

Bitopertin, MedChemExpress

Product Code. 16482976
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482976 25 mg 25mg
16465136 100 mg 100mg
16444178 10 mg 10mg
16440527 50 mg 50mg
16406887 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16482976 Supplier Medchem Express Supplier No. HY10809

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bitopertin is a potent, noncompetitive glycine reuptake inhibitor, inhibits glycine uptake at human GlyT1 with a concentration exhibiting IC50 of 25 nM.

Bitopertin is a potent, noncompetitive glycine reuptake inhibitor, inhibits glycine uptake at human GlyT1 with a concentration exhibiting IC50 of 25 nM.

Specifications

Chemical Name or Material Bitopertin
CAS 845614-11-1
Color White
Purity Grade Notes Research
Molecular Formula C21H20F7N3O4S
Quantity 25 mg
Synonym RG1678 RO4917838
Solubility Information DMSO : ≥ 50 mg/mL (92.00 mM)
SMILES FC1=CC(C(F)(F)F)=CN=C1N2CCN(C(C3=CC(S(=O)(C)=O)=CC=C3O[C@H](C(F)(F)F)C)=O)CC2
Molecular Weight (g/mol) 543.46
Formula Weight 543.46
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.