missing translation for 'onlineSavingsMsg'
Learn More

Bithionol sulfoxide, MedChemExpress

Product Code. 16481886
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 16481886

Brand: Medchem Express HY17592A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bithionol sulfoxide(Bitin-S) is a clinically approved anti-parasitic drug; has been shown to inhibit solid tumor growth in several preclinical cancer models.

Bithionol sulfoxide(Bitin-S) is a clinically approved anti-parasitic drug

Specifications

Chemical Name or Material Bithionol sulfoxide
Physical Form Solid
CAS 844-26-8
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C12H6Cl4O3S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 250 mg/mL (671.95 mM)
SMILES O=S(C1=CC(Cl)=CC(Cl)=C1O)C2=CC(Cl)=CC(Cl)=C2O
Molecular Weight (g/mol) 372.05
Formula Weight 372.05
Percent Purity 98.39%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.