missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bisphosphoglycerate mutase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553434
This item is not returnable.
View return policy
Description
Immunogen sequence: RRSYNVTPPP IEESHPYYQE IYNDRRYKVC DVPLDQLPRS ESLKDVLERL LPYWNERIAP EVLRGKTILI SAHGNSSRAL LKHLEGISDE DIINITLPTG VPILLELDEN LRAVGPHQFL GDQEAIQAAI KKVEDQGKVK QA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
Riboflavin kinase is an essential enzyme that catalyzes the phosphorylation of riboflavin to form flavin mononucleotide, an obligatory step in vitamin B2 utilization and flavin cofactor synthesis.
Specifications
| Bisphosphoglycerate mutase | |
| Polyclonal | |
| Unconjugated | |
| BPGM | |
| 2,3-bisphosphoglycerate mutase; 2,3-bisphosphoglycerate mutase, erythrocyte; 2,3-bisphosphoglycerate synthase; 2,3-diphosphoglycerate mutase; Ab2-098; AI323730; AL022789; Bisphosphoglycerate mutase; BPG-dependent PGAM; Bpgm; C86192; DPGM; erythrocyte 2,3-bisphosphoglycerate mutase; hypothetical protein LOC533785; testis secretory sperm-binding protein Li 202a; zgc:92230 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12183, 296973, 669 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07738, P15327 | |
| BPGM | |
| Recombinant protein corresponding to Human Bisphosphoglycerate mutase. Recombinant protein control fragment (Product #RP-90910). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction