Learn More
Bisphosphoglycerate mutase Polyclonal Antibody
GREENER_CHOICE
Bisphosphoglycerate mutase Polyclonal Antibody
Brand:  Invitrogen PA544868
Product Code. 15576672
  • £442.00 / 100µL
Description
Bisphosphoglycerate mutase Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bisphosphoglycerate mutase
Polyclonal
Unconjugated
BPGM
2,3-bisphosphoglycerate mutase; 2,3-bisphosphoglycerate mutase, erythrocyte; 2,3-bisphosphoglycerate synthase; 2,3-diphosphoglycerate mutase; Ab2-098; AI323730; AL022789; Bisphosphoglycerate mutase; BPG-dependent PGAM; Bpgm; C86192; DPGM; erythrocyte 2,3-bisphosphoglycerate mutase; hypothetical protein LOC533785; testis secretory sperm-binding protein Li 202a; zgc:92230
Rabbit
Affinity Chromatography
RUO
669
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P07738
BPGM
Synthetic peptide directed towards the middle region of human BPGM.