missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol S Monosulfate Disodium Salt, TRC

Product Code. 30137080
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137080 25 mg 25mg
30009891 5 mg 5mg
30011164 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137080 Supplier Toronto Research Chemicals Supplier No. TRCB51962825MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12H8Na2O7S2
Molecular Weight (g/mol) 374.3
Synonym 4-((4-Hydroxyphenyl)sulfonyl)phenyl Hydrogen Sulfate Disodium Salt, BPS Monosulfate Disodium Salt;
IUPAC Name disodium;[4-(4-oxidophenyl)sulfonylphenyl] sulfate
SMILES O=S(OC(C=C1)=CC=C1S(C2=CC=C(O[Na])C=C2)(=O)=O)(O[Na])=O

Specifications

Analyte or Component Names Bisphenol S Monosulfate Disodium Salt
Shipping Condition Room Temperature
InChI Formula InChI=1S/C12H10O7S2.2Na/c13-9-1-5-11(6-2-9)20(14,15)12-7-3-10(4-8-12)19-21(16,17)18;;/h1-8,13H,(H,16,17,18);;/q;2*+1/p-2
Solution Type Neat
Industry Type Pharmaceutical
Quantity 25 mg
Percent Purity >95%
Chemical Name or Material Bisphenol S Monosulfate Disodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.