missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A, TRC

Product Code. 30040354
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040354 10 g 10g
17919629 100 mg 100mg
18043310 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040354 Supplier Toronto Research Chemicals Supplier No. TRCB51949510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

Chemical Identifiers

CAS 80-05-7
Molecular Formula C15 H16 O2
Molecular Weight (g/mol) 228.29
Synonym Phenol, 4,4'-isopropylidenedi- (8CI), 4,4'-(1-Methylethylidene)bis[phenol], (4,4'-Dihydroxydiphenyl)dimethylmethane, 2,2-Bis(4-hydroxyphenyl)propane, 2,2-Bis(p-hydroxyphenyl)propane, 2,2-Di(4-hydroxyphenyl)propane, 2,2-Di(4-phenylol)propane, 2,2'-Bis(4-hydroxyphenyl)propane, 4,4'-(Propane-2,2-diyl)diphenol, 4,4'-Isopropylidenebis[phenol], 4,4'-Isopropylidenediphenol, 4,4'-Methylethylidenebisphenol, B 0494, BPA, BPA 154, BPA 157, BPA-M, Bis(4-hydroxyphenyl)dimethylmethane, Bis(p-hydroxyphenyl)propane, Bisphenol A, Dian, Diano, Diphenylolpropane, HT 3082, Hidorin F 285, Hidorin F 568, Ipognox 88, Isopropylidenebis(4-hydroxybenzene), NSC 1767, NSC 17959, Parabis, Parabis A, Pluracol 245, Rikabanol, p,p'-Bisphenol A, p,p'-Dihydroxydiphenylpropane, p,p'-Isopropylidenebisphenol, p,p'-Isopropylidenediphenol, β,β'-Bis(p-hydroxyphenyl)propane
IUPAC Name 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
SMILES CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3
Formula Weight 228.115
Percent Purity >95%
Chemical Name or Material Bisphenol A
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.