missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A bis(4-aminophenyl) ether, Dr. Ehrenstorfer

Product Code. 30324411
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324411 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324411 Supplier Dr. Ehrenstorfer Supplier No. DREC10655525
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bisphenol A Bis(4-aminophenyl) Ether
CAS 13080-86-9
Analyte or Component Names Bisphenol A Bis(4-aminophenyl) Ether
Shipping Condition +20°C
Molecular Formula C27 H26 N2 O2
Certifications/Compliance ISO/IEC 17025
Synonym 4,4'-[(1-Methylethylidene)bis(4,1-phenyleneoxy)]bis[benzenamine] (ACI), Aniline, 4,4'-[isopropylidenebis(p-phenyleneoxy)]di- (7CI, 8CI), 2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 2,2-Bis[p-(4-aminophenoxy)phenyl]propane, 4,4'-(4,4'-Isopropylidenediphenyl-1,1'-diyldioxy)dianiline, 4,4'-[Isopropylidenebis(1,4-phenylene)dioxy]dianiline, 4,4'-Propane-2,2-diylbis(4,1-phenyleneoxy)dianiline, 4-[4-[2-[4-(4-Aminophenoxy)phenyl]propan-2-yl]phenoxy]benzenamine, [4-(4-[1-[4-(4-Aminophenoxy)phenyl]-1-methylethyl]phenoxy)phenyl]amine, BAPP, Bisphenol A bis(4-aminophenyl) ether
InChI Formula InChI=1S/C27H26N2O2/c1-27(2,19-3-11-23(12-4-19)30-25-15-7-21(28)8-16-25)20-5-13-24(14-6-20)31-26-17-9-22(29)10-18-26/h3-18H,28-29H2,1-2H3
SMILES CC(C)(c1ccc(Oc2ccc(N)cc2)cc1)c3ccc(Oc4ccc(N)cc4)cc3
IUPAC Name 4-[4-[2-[4-(4-aminophenoxy)phenyl]propan-2-yl]phenoxy]aniline
Molecular Weight (g/mol) 410.507
Quantity 10 mg
Formula Weight 410.199 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.