missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A acetate propionate, epoxy curative, EC-326, Thermo Scientific™

Product Code. 15437215
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Recommended for use as an epoxy curative to improve toughness, hydrophobicity, and hydrolytic stability
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071466-57-3
Molecular Formula C20H22O4
Molecular Weight (g/mol) 326.392
MDL Number MFCD22380701
InChI Key VWMUCEDXVAQLNS-UHFFFAOYSA-N
Synonym bisphenol a acetate propionate, technical grade, bisphenol a acetate propionate, epoxy curative, ec-326, 4-2-4-acetyloxy phenyl propan-2-yl phenyl propanoate
PubChem CID 59317407
IUPAC Name [4-[2-(4-acetyloxyphenyl)propan-2-yl]phenyl] propanoate
SMILES CCC(=O)OC1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)OC(=O)C

Specifications

Color White to Yellow
Physical Form White to yellow solid
Quantity 25 g
Formula Weight 326.39
Chemical Name or Material Bisphenol A acetate propionate, Epoxy curative, EC-326
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'