missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A, 97+%

Product Code. 11444093
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444093 1000 g 1000g
11454093 250 g 250g
11464093 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444093 Supplier Thermo Scientific Alfa Aesar Supplier No. A10324.0B
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-05-7 | C15H16O2 | 228.29 g/mol

Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).

Solubility
Soluble in aqueous alkaline solution, acetic acid, alcohol and acetone. Slightly soluble in carbon tetrachloride. It has poor solubility in water.

Notes
Incompatible with acid anhydrides, acid chlorides, strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-05-7
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00002366
InChI Key IISBACLAFKSPIT-UHFFFAOYSA-N
Synonym bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian
PubChem CID 6623
ChEBI CHEBI:33216
IUPAC Name 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
SMILES CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 153°C to 158°C
Boiling Point 200°C (4 mmHg)
Flash Point 227°C (440°F)
Quantity 1000 g
Beilstein 1107700
Merck Index 14,1297
Solubility Information Soluble in aqueous alkaline solution,acetic acid,alcohol and acetone. Slightly soluble in carbon tetrachloride. It has poor solubility in water.
Formula Weight 228.29
Percent Purity ≥97%
Chemical Name or Material Bisphenol A
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bisphenol A
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H360F-May damage fertility.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Bisphenol A

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.