missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A, 97+%, Thermo Scientific Chemicals

Catalog Number p-7080688
Click to view available options
Quantity:
250 g
1000 g
5000 g
80-05-7
C15H16O2
228.29
MFCD00002366
IISBACLAFKSPIT-UHFFFAOYSA-N
bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian
6623
CHEBI:33216
4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
80-05-7
C15H16O2
228.29
MFCD00002366
IISBACLAFKSPIT-UHFFFAOYSA-N
bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian
6623
CHEBI:33216
4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).

Solubility
Soluble in aqueous alkaline solution, acetic acid, alcohol and acetone. Slightly soluble in carbon tetrachloride. It has poor solubility in water.

Notes
Incompatible with acid anhydrides, acid chlorides, strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 153°C to 158°C
Boiling Point 200°C (4 mmHg)
Flash Point 227°C (440°F)
Quantity 250 g
Beilstein 1107700
Merck Index 14,1297
Solubility Information Soluble in aqueous alkaline solution,acetic acid,alcohol and acetone. Slightly soluble in carbon tetrachloride. It has poor solubility in water.
Formula Weight 228.29
Percent Purity ≥97%
Chemical Name or Material Bisphenol A

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'